Assay Details
| Gene Symbol: 4930548K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040656.1 | 2 - 3 | 290 | 180 | ||
| GenBank mRNA | AK016071.1 | - | 2 - 3 | 290 | 180 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
mannosidase, endo-alpha |
Manea | 4932703L02Rik | -289.2kb | Sense | Chr. 4 - 26324506 - 26346652 on NCBI Build GRCm38 |
| Downstream | Eph receptor A7 | Epha7 |
Cek11, Ebk, Ehk3, Hek11, Mdk1 |
2.1mb | Anti-sense | Chr. 4 - 28813044 - 28967511 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0500002443.rn
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 959290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784563 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491600 | AK049194.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491600 | AK050045.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491600 | AK076793.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491600 | AK168103.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491600 | AK168534.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491600 | AK170510.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491600 | AK176998.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491600 | AK205803.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491600 | AK218145.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491600 | NM_133728.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350302 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447337 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 253212 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703317 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 255366 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495228 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147816 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727055 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 5591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536384 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214984 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745504 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193760 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86219 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 21571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120416 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05183722 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05183701 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05183650 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05183680 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05183722 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05183650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04012283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03829003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03522415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03473730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03522431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03990048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03231470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03305542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03522432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03231485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22794637 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0400019142.mm.1 | NR_040656.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400019142.mm.1 | NR_040656.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400019142.mm.1 | TC0400002335.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400019141.mm.1 | NR_040656.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400019141.mm.1 | NR_040656.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400019141.mm.1 | TC0400002335.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400019143.mm.1 | NR_040656.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400019143.mm.1 | NR_040656.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400019143.mm.1 | TC0400002335.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 306706 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100630044 | - | - |

4930548K13Rik
Genomic Map