Assay Details
| Gene Symbol: 4930551I15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016092.1 | - | 1 | 949 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | inositol polyphosphate multikinase | Ipmk |
2410017C19Rik, AA408208, Impk |
-98.4kb | Anti-sense | Chr. 10 - 71347765 - 71385885 on NCBI Build GRCm38 |
| Downstream | hematological and neurological expressed 1-like pseudogene | 1700049L16Rik | 494.4kb | Sense | Chr. 10 - 71979885 - 71980690 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930551I15Rik
Genomic Map