Assay Details
| Gene Symbol: 4930552N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040661.1 | 1 - 2 | 133 | 116 | ||
| GenBank mRNA | AK016098.1 | - | 1 - 2 | 134 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
topoisomerase I binding, arginine/serine-rich like |
Toporsl | 4930547C10Rik | -202.2kb | Sense | Chr. 4 - 52596274 - 52612424 on NCBI Build GRCm38 |
| Downstream | olfactory receptor 275 | Olfr275 | MOR262-2 | 10.0kb | Sense | Chr. 4 - 52825399 - 52826358 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930552N02Rik
Genomic Map