Assay Details
| Gene Symbol: 4930553C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016100.1 | - | 1 - 2 | 582 | 141 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 12130 | Gm12130 | OTTMUSG00000005424 | -867.4kb | Sense | Chr. 11 - 38491548 - 38520045 on NCBI Build GRCm38 |
| Downstream | predicted gene 12132 | Gm12132 | OTTMUSG00000005426 | 618.0kb | Anti-sense | Chr. 11 - 40008194 - 40337842 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930553C11Rik
Genomic Map