Assay Details
| Gene Symbol: 4930560O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016186.1 | - | 1 | 1050 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 159 | Tmem159 | 8430420C20Rik | Anti-sense | Chr. 7 - 120102426 - 120120987 on NCBI Build GRCm38 | |
| Downstream | zona pellucida glycoprotein 2 | Zp2 | Zp-2 | 23.6kb | Sense | Chr. 7 - 120126772 - 120145754 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930560O18Rik
Genomic Map