Assay Details
| Gene Symbol: 4930572C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016277.1 | - | 1 | 1050 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | giant axonal neuropathy | Gan |
A330045G18, gigaxonin |
-85.5kb | Sense | Chr. 8 - 117158135 - 117205186 on NCBI Build GRCm38 |
| Downstream | microRNA 7077 | Mir7077 |
Gm27774, mmu-mir-7077 |
160.8kb | Sense | Chr. 8 - 117453023 - 117453079 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930572C08Rik
Genomic Map