Assay Details
| Gene Symbol: 4930578M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016308.1 | - | 1 - 2 | 319 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | THAP domain containing 11 | Thap11 |
2810036E22Rik, AB041579, CTG-B43a, CTG-B45d, Ronin |
-2.5kb | Anti-sense | Chr. 8 - 105855103 - 105856950 on NCBI Build GRCm38 |
| Downstream | nuclear transport factor 2 | Nutf2 |
2700067I02Rik, AI115459, AI413596, AW546000, Ntf2, Ntf3, PP15 |
212.0 | Anti-sense | Chr. 8 - 105860634 - 105880402 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930578M07Rik
Genomic Map