Assay Details
| Gene Symbol: 4930579K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029444.1 | 1 | 80 | 85 | ||
| GenBank mRNA | AK016329.1 | - | 1 | 53 | 85 | |
| AK077474.1 | - | 1 | 85 | |||
| AK087620.1 | - | 1 | 81 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
retinol binding protein 2, cellular |
Rbp2 |
Crbp-2, CrbpII, Rbp-2 |
-52.7kb | Anti-sense | Chr. 9 - 98490537 - 98509771 on NCBI Build GRCm38 |
| Downstream |
coatomer protein complex, subunit beta 2 (beta prime) |
Copb2 | AI256832 | 148.0 | Anti-sense | Chr. 9 - 98563731 - 98588375 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077474.1 transcripts.
Set Membership: |
|

4930579K19Rik
Genomic Map