Assay Details
| Gene Symbol: Metap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016346.1 | - | 2 - 3 | 225 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin specific peptidase 44 | Usp44 | E430004F17Rik | -29.3kb | Anti-sense | Chr. 10 - 93819968 - 93860738 on NCBI Build GRCm38 |
| Downstream |
vezatin, adherens junctions transmembrane protein |
Vezt |
A630071D13Rik, AI848282 |
70.3kb | Sense | Chr. 10 - 93961522 - 94035799 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Metap2
Genomic Map