Assay Details
| Gene Symbol: 4930588G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016366.1 | - | 3 - 4 | 1180 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PDZ domain containing 9 | Pdzd9 | 4930408O21Rik | Anti-sense | Chr. 7 - 120659104 - 120677562 on NCBI Build GRCm38 | |
| Downstream | cDNA sequence BC030336 | BC030336 | 1.7kb | Sense | Chr. 7 - 120677620 - 120734854 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930588G17Rik
Genomic Map