Assay Details
| Gene Symbol: 4930597E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016404.1 | - | 1 | 552 | 119 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
non-SMC condensin II complex, subunit H2 pseudogene |
Gm8363 | EG666919 | -260.3kb | Sense | Chr. 17 - 5480989 - 5483069 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 3300005D01 gene | 3300005D01Rik | 54.4kb | Anti-sense | Chr. 17 - 5798657 - 5803242 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930597E08Rik
Genomic Map