Assay Details
| Gene Symbol: 4931407J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016442.1 | - | 1 | 1350 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 690 | Mir690 |
Mirn690, mir-690, mmu-mir-690 |
-30.5kb | Sense | Chr. 16 - 28599935 - 28600043 on NCBI Build GRCm38 |
| Downstream | Mab-21 domain containing 2 | Mb21d2 |
1600021P15Rik, A430031N04, C87006 |
194.1kb | Sense | Chr. 16 - 28826176 - 28929698 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4931407J08Rik
Genomic Map