Assay Details
| Gene Symbol: 4931412I15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016451.1 | - | 1 | 1248 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
discs, large homolog 2 (Drosophila) |
Dlg2 |
A330103J02Rik, B230218P12Rik, B330007M19Rik, Dlgh2, Gm1197, PSD93 |
Anti-sense | Chr. 7 - 90476183 - 92449246 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930567K12 gene | 4930567K12Rik | 10.0kb | Sense | Chr. 7 - 92092554 - 92108692 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4931412I15Rik
Genomic Map