Assay Details
| Gene Symbol: 4932437C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016552.1 | - | 1 | 2349 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | C-terminal binding protein 2 | Ctbp2 |
AA407280, D7Ertd45e, Gtrgeo6, Ribeye |
Anti-sense | Chr. 7 - 132986417 - 133123877 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930483O08 gene | 4930483O08Rik | 121.4kb | Sense | Chr. 7 - 133128251 - 133142457 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4932437C15Rik
Genomic Map