Assay Details
| Gene Symbol: 4932702M13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016579.1 | - | 1 | 2650 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kelch-like 32 | Klhl32 |
6430524H05Rik, D4Ertd389e, Gm1356, mKIAA1900 |
-43.8kb | Sense | Chr. 4 - 24614352 - 24851110 on NCBI Build GRCm38 |
| Downstream |
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 4 |
Ndufaf4 |
1110007M04Rik, 3000003G13Rik, AW214064 |
108.0 | Anti-sense | Chr. 4 - 24898083 - 24905001 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4932702M13Rik
Genomic Map