Assay Details
| Gene Symbol: 2610209C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016706.1 | - | 1 | 854 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc and ring finger 2 | Znrf2 |
1190002C14Rik, AU044205, D6Ertd365e |
Anti-sense | Chr. 6 - 54816916 - 54890224 on NCBI Build GRCm38 | |
| Downstream | nucleotide-binding oligomerization domain containing 1 | Nod1 |
C230079P11, Card4, F830007N14Rik, Nlrc1 |
107.0kb | Sense | Chr. 6 - 54923942 - 54972672 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2610209C05Rik
Genomic Map