Assay Details
| Gene Symbol: 4933407I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016723.1 | - | 1 | 1256 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6980 | Gm6980 | Anti-sense | Chr. 9 - 51915912 - 51930268 on NCBI Build GRCm38 | ||
| Downstream | ferredoxin 1 | Fdx1 | 27.0kb | Anti-sense | Chr. 9 - 51943023 - 51963602 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933407I05Rik
Genomic Map