Assay Details
| Gene Symbol: 4933407O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016733.1 | - | 1 | 947 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heterogeneous nuclear ribonucleoprotein L-like | Hnrnpll |
2510028H02Rik, 2810036L13Rik, AI256697, AI852082, Hnrpll |
-288.0 | Anti-sense | Chr. 17 - 80029487 - 80062513 on NCBI Build GRCm38 |
| Downstream | galactose mutarotase | Galm |
A530057M15Rik, AU015645, AU020959 |
63.2kb | Sense | Chr. 17 - 80127458 - 80185032 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933407O12Rik
Genomic Map