Assay Details
| Gene Symbol: 4933421H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016863.1 | - | 1 | 1449 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hematopoietically expressed homeobox | Hhex |
Hex, Hex1, Hhex-rs2, Prh, Prhx |
-24.5kb | Sense | Chr. 19 - 37434841 - 37440731 on NCBI Build GRCm38 |
| Downstream | exocyst complex component 6 | Exoc6 |
4833405E05Rik, AW413330, C430002C19, Sec15, Sec15l1, hbd, msec15 |
53.9kb | Sense | Chr. 19 - 37520874 - 37684052 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933421H12Rik
Genomic Map