Assay Details
| Gene Symbol: 4933423N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016877.1 | - | 1 | 1054 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | endothelial PAS domain protein 1 | Epas1 |
HIF-2alpha, HIF2A, HLF, HRF, MOP2, bHLHe73 |
-82.2kb | Anti-sense | Chr. 17 - 86753864 - 86833410 on NCBI Build GRCm38 |
| Downstream | transmembrane protein 247 | Tmem247 | 1700090G07Rik | 88.0 | Anti-sense | Chr. 17 - 86917348 - 86922482 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933423N03Rik
Genomic Map