Assay Details
| Gene Symbol: 4933424C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016885.1 | - | 1 | 1450 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin carboxy-terminal hydrolase L1 | Uchl1 |
AW822034, C88048, PGP 9.5, PGP9.5, R75593, UCH-L1, UCHL-1, gad |
-31.1kb | Anti-sense | Chr. 5 - 66676121 - 66687234 on NCBI Build GRCm38 |
| Downstream | LIM and calponin homology domains 1 | Limch1 |
3732412D22Rik, mKIAA1102 |
25.9kb | Anti-sense | Chr. 5 - 66745840 - 67057159 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115039 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05600402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04994936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04111920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04497805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04111939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04711339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04111918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04327914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04711343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04292995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04984964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04111934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04607802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04607805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23763531 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1454300_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432617_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454300_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454300_at | - | - |
| Mouse Genome 430 2.0 Array | 1432617_at | - | - |
| Mouse Genome 430 2.0 Array | 1432617_PM_at | - | - |
| Mouse Expression Set 430 | 1432617_at | - | - |
| Mouse Expression Set 430 | 1454300_PM_at | - | - |
| Mouse Expression Set 430 | 1432617_PM_at | - | - |
| Mouse Expression Set 430 | 1454300_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432617_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454300_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454300_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432617_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 503259 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101500048 | - | - |

4933424C09Rik
Genomic Map