Assay Details
| Gene Symbol: 4933424H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016894.1 | - | 1 | 1150 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA D030025E07 gene | D030025E07Rik | -467.9kb | Sense | Chr. 3 - 128117015 - 128231605 on NCBI Build GRCm38 | |
| Downstream | paired-like homeodomain transcription factor 2 | Pitx2 |
9430085M16Rik, Brx1, Brx1b, Munc30, Otlx2, Ptx2, Rieg |
498.3kb | Anti-sense | Chr. 3 - 129199878 - 129219594 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933424H11Rik
Genomic Map