Assay Details
| Gene Symbol: 4933426I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016932.1 | - | 1 | 254 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 938 | Zfp938 | B230315N10Rik | -74.1kb | Sense | Chr. 10 - 82224845 - 82241318 on NCBI Build GRCm38 |
| Downstream | predicted gene 4924 | Gm4924 |
EG237412, Mif1 |
37.7kb | Anti-sense | Chr. 10 - 82354237 - 82379643 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933426I03Rik
Genomic Map