Assay Details
| Gene Symbol: 4933431C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017001.1 | - | 1 | 448 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
NLR family, pyrin domain containing 4C |
Nlrp4c |
Nalp-alpha, Nalp4c, Rnh2 |
-30.9kb | Sense | Chr. 7 - 6045161 - 6105509 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 444 | Zfp444 |
2810031J10Rik, 6230401O10Rik, mFLJ00134 |
34.8kb | Sense | Chr. 7 - 6172462 - 6193104 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933431C10Rik
Genomic Map