Assay Details
| Gene Symbol: 4933431K14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017009.1 | - | 1242 | 88 | ||
| BB762387.1 | - | 432 | 88 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 46, member A |
Fam46a | D930050G01Rik | -423.0kb | Sense | Chr. 9 - 85320439 - 85327150 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 9330154J02 gene | 9330154J02Rik | 70.1kb | Sense | Chr. 9 - 85820490 - 85842955 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933431K14Rik
Genomic Map