Assay Details
| Gene Symbol: 4930533K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017042.1 | - | 1 - 2 | 169 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA D630013N20 gene | D630013N20Rik | -212.7kb | Anti-sense | Chr. 10 - 70601049 - 70655963 on NCBI Build GRCm38 | |
| Downstream | bicaudal C homolog 1 (Drosophila) | Bicc1 |
Bic-C, bpk, jcpk |
52.1kb | Anti-sense | Chr. 10 - 70922837 - 71159746 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930533K18Rik
Genomic Map