Assay Details
| Gene Symbol: 4933435G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017069.1 | - | 1 | 1649 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6567 | Gm6567 | -22.6kb | Anti-sense | Chr. 7 - 73097752 - 73102068 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 20083 |
Gm20083 | 57.1kb | Sense | Chr. 7 - 73183605 - 73192432 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2212344 | AK017069.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06300448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06168479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06524467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06407939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05994182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06189563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06613100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06613088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06613104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06125594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06125610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06168481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06189580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05933004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06409006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06008027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06008025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06189565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06309196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06613101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06014384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06428051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06407937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06300451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06641441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06168478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25227235 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1432832_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432833_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432833_at | - | - |
| Mouse Genome 430 2.0 Array | 1432832_at | - | - |
| Mouse Genome 430 2.0 Array | 1432832_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432833_PM_at | - | - |
| Mouse Expression Set 430 | 1432832_PM_at | - | - |
| Mouse Expression Set 430 | 1432832_at | - | - |
| Mouse Expression Set 430 | 1432833_at | - | - |
| Mouse Expression Set 430 | 1432833_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432832_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432833_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432832_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432833_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 412202 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103990270 | - | - |

4933435G04Rik
Genomic Map