Assay Details
| Gene Symbol: 4933436P19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017091.1 | - | 1 | 1151 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
pyridoxal (pyridoxine, vitamin B6) kinase |
Pdxk |
2310036D04Rik, AA119688, C77999 |
-4.9kb | Sense | Chr. 10 - 78436747 - 78464948 on NCBI Build GRCm38 |
| Downstream | ilvB (bacterial acetolactate synthase)-like | Ilvbl |
5830463I21, AI415009 |
103.1kb | Anti-sense | Chr. 10 - 78574345 - 78584502 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933436P19Rik
Genomic Map