Assay Details
| Gene Symbol: 4933437I04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017101.1 | - | 1 | 350 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
poly(A) binding protein, cytoplasmic 1-like |
Pabpc1l |
1810053B01Rik, Epab |
-10.3kb | Sense | Chr. 2 - 164024770 - 164050538 on NCBI Build GRCm38 |
| Downstream | serine/threonine kinase 4 | Stk4 |
AI447339, AU020804, Kas-2, Mst1, Ysk3 |
11.8kb | Sense | Chr. 2 - 164074109 - 164155524 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933437I04Rik
Genomic Map