Assay Details
| Gene Symbol: 5031415H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017142.1 | - | 1 | 1148 | 102 | |
| AK081553.1 | - | 1 | 1821 | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 36594 |
Gm36594 | -854.0kb | Anti-sense | Chr. 17 - 69894630 - 69897331 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 41609 |
Gm41609 | 13.5kb | Anti-sense | Chr. 17 - 70766096 - 70782793 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5031415H12Rik
Genomic Map