Assay Details
| Gene Symbol: Itgb2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017157.1 | - | 1 | 1352 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 5 |
B3galt5 |
1190002B21Rik, AU045265, b3Galt-V |
-111.5kb | Sense | Chr. 16 - 96235801 - 96319859 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 32358 |
Gm32358 | 760.0 | Sense | Chr. 16 - 96433632 - 96447014 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Itgb2l
Genomic Map