Assay Details
| Gene Symbol: 5033428C03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017200.1 | - | 1 | 1246 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | amphiregulin | Areg |
AR, Mcub, Sdgf |
-61.0kb | Sense | Chr. 5 - 91139615 - 91148432 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 19619 |
Gm19619 | 46.7kb | Anti-sense | Chr. 5 - 91257571 - 91283076 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5033428C03Rik
Genomic Map