Assay Details
| Gene Symbol: 5430404G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017264.1 | - | 1 | 1954 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | abhydrolase domain containing 10 | Abhd10 | -20.4kb | Sense | Chr. 16 - 45729720 - 45743278 on NCBI Build GRCm38 | |
| Downstream | predicted gene 15638 | Gm15638 | OTTMUSG00000024690 | 178.6kb | Anti-sense | Chr. 16 - 45944419 - 45957495 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5430404G13Rik
Genomic Map