Assay Details
| Gene Symbol: Clcn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017285.1 | - | 1 | 849 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | A kinase (PRKA) anchor protein 4 | Akap4 |
AKAP-4, AKAP82, Fsc1, PRKA4, mAKAP82, p82 |
-75.2kb | Anti-sense | Chr. X - 7067515 - 7078609 on NCBI Build GRCm38 |
| Downstream | microRNA 500 | Mir500 |
Mirn500, mir-500, mmu-mir-500 |
82.5kb | Sense | Chr. X - 7237683 - 7237774 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Clcn5
Genomic Map