Assay Details
| Gene Symbol: 5430410E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017289.1 | - | 1 | 1154 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B | Ddx39b |
0610030D10Rik, AI428441, Bat-1, Bat1, Bat1a, D17H6S81E, D17H6S81E-1, D6S81Eh |
-8.6kb | Anti-sense | Chr. 17 - 35241746 - 35253707 on NCBI Build GRCm38 |
| Downstream |
histocompatibility 2, Q region locus 1 |
H2-Q1 |
H-2Q1, Q1, Q1b, Q1d, Q1k, Qa-1, Qa1, Qed-1 |
56.9kb | Anti-sense | Chr. 17 - 35320516 - 35327699 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5430410E06Rik
Genomic Map