Assay Details
| Gene Symbol: Ammecr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242430.1 | NP_001229359.1 | 8 | 4328 | 74 | |
| NM_153515.4 | NP_705735.1 | 8 | 4127 | 74 | ||
| GenBank mRNA | AK017359.1 | - | 1 | 849 | 74 | |
| AK049081.1 | - | 1 | 916 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Sin3A associated protein | Sap130 |
2610304F09Rik, 6720406D06 |
-59.5kb | Sense | Chr. 18 - 31634327 - 31723068 on NCBI Build GRCm38 |
| Downstream | polymerase (RNA) II (DNA directed) polypeptide D | Polr2d |
2310002G05Rik, 2610028L19Rik, HSRBP4, RBP4 |
5.4kb | Sense | Chr. 18 - 31789159 - 31796701 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ammecr1l
Genomic Map