Assay Details
| Gene Symbol: 5430430K15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017366.1 | - | 1 | 746 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | matrix metallopeptidase 14 (membrane-inserted) | Mmp14 |
AI325305, MT-MMP-1, MT1-MMP, sabe |
-5.1kb | Sense | Chr. 14 - 54431604 - 54441258 on NCBI Build GRCm38 |
| Downstream | low-density lipoprotein receptor-related protein 10 | Lrp10 | Lrp9 | 16.3kb | Sense | Chr. 14 - 54464147 - 54470292 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5430430K15Rik
Genomic Map