Assay Details
| Gene Symbol: Cpeb1os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017413.1 | - | 1 | 52 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytoplasmic polyadenylation element binding protein 1 | Cpeb1 |
AU024112, Cpeb, mCPEB |
-438.0 | Anti-sense | Chr. 7 - 81347026 - 81455457 on NCBI Build GRCm38 |
| Downstream |
adaptor-related protein complex 3, beta 2 subunit |
Ap3b2 |
AI549966, AU042881, Naptb, [b]-NAP, beta-NAP, beta3B |
2.4kb | Anti-sense | Chr. 7 - 81460399 - 81494038 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cpeb1os1
Genomic Map