Assay Details
| Gene Symbol: 5730405O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017499.1 | - | 1 | 750 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosomal protein S24 | Mrps24 |
3110030K20Rik, AI414579, Rpms24 |
-49.2kb | Sense | Chr. 11 - 5703983 - 5707717 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2210015D19 gene | 2210015D19Rik |
1600028I17Rik, AI596465 |
3.9kb | Anti-sense | Chr. 11 - 5762150 - 5784710 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5730405O12Rik
Genomic Map