Assay Details
| Gene Symbol: 5730409L17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017536.1 | - | 1 | 851 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | islet cell autoantigen 1 | Ica1 |
69kDa, ICA69 |
Sense | Chr. 6 - 8630527 - 8778537 on NCBI Build GRCm38 | |
| Downstream | neurexophilin 1 | Nxph1 | C130005L03Rik | 219.9kb | Anti-sense | Chr. 6 - 8948614 - 9259863 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5730409L17Rik
Genomic Map