Assay Details
| Gene Symbol: 5730419A17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017574.1 | - | 1 | 543 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 120, member C |
Fam120c |
D930001I21Rik, ORF34 |
-68.2kb | Sense | Chr. X - 151342932 - 151474136 on NCBI Build GRCm38 |
| Downstream |
HECT, UBA and WWE domain containing 1 |
Huwe1 |
5430439H10Rik, AU041296, Arf-bp1, C430014N20Rik, C80292, Gm1718, Ib772, LASU1, Mule, Ureb1 |
257.7kb | Sense | Chr. X - 151800807 - 151935417 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5730419A17Rik
Genomic Map