Assay Details
| Gene Symbol: 5730458M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017674.1 | - | 1 | 1555 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CD302 antigen | Cd302 |
1110055L24Rik, AI159627 |
Anti-sense | Chr. 2 - 60251993 - 60284484 on NCBI Build GRCm38 | |
| Downstream | lymphocyte antigen 75 | Ly75 |
CD205, DEC-205, DEC205 |
38.4kb | Anti-sense | Chr. 2 - 60293760 - 60383231 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5730458M16Rik
Genomic Map