Assay Details
| Gene Symbol: 5730478J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017704.1 | - | 1 | 1355 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | methyl-CpG binding domain protein 2 | Mbd2 | MBD2a | -1.5mb | Anti-sense | Chr. 18 - 70568270 - 70626131 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 31819 |
Gm31819 | 527.8kb | Sense | Chr. 18 - 72686426 - 72725083 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5730478J17Rik
Genomic Map