Assay Details
| Gene Symbol: 5830415G21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017932.1 | - | 1 | 1248 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6209 | Gm6209 | EG621304 | Sense | Chr. 3 - 50516399 - 50628670 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 37548 |
Gm37548 | 306.7kb | Sense | Chr. 3 - 50883237 - 50888892 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5830415G21Rik
Genomic Map