Assay Details
| Gene Symbol: 5830445D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017990.1 | - | 1 | 754 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L10 pseudogene | D11Bhm181e | -53.9kb | Sense | Chr. 11 - 78561311 - 78561600 on NCBI Build GRCm38 | |
| Downstream |
family with sequence similarity 58, member B |
Fam58b |
1810009O10Rik, Fam58a |
133.9kb | Sense | Chr. 11 - 78750506 - 78751729 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5830445D09Rik
Genomic Map