Assay Details
| Gene Symbol: Amn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018037.1 | - | 1 | 1353 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | methyltransferase like 20 | Mettl20 |
4833442J19Rik, AI256744 |
-31.8kb | Anti-sense | Chr. 6 - 149141513 - 149151171 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2810474O19 gene | 2810474O19Rik |
6720435I21Rik, AA536804, AU020969, GET, Kiaa1551 |
124.7kb | Anti-sense | Chr. 6 - 149309414 - 149335663 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Amn1
Genomic Map