Assay Details
| Gene Symbol: Edaradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018047.1 | - | 1 | 1149 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
lectin, galactose binding, soluble 8 |
Lgals8 |
1200015E08Rik, AI326142, D13Ertd524e, Lgals-8 |
-53.8kb | Sense | Chr. 13 - 12439402 - 12464976 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 30239 |
Gm30239 | 19.8kb | Anti-sense | Chr. 13 - 12539888 - 12562716 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Edaradd
Genomic Map