Assay Details
| Gene Symbol: 6330419E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018200.1 | - | 1 | 1050 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thiamine pyrophosphokinase | Tpk1 | -1.6mb | Anti-sense | Chr. 6 - 43344999 - 43666484 on NCBI Build GRCm38 | |
| Downstream | cullin 1 | Cul1 | 2.1mb | Sense | Chr. 6 - 47452273 - 47526139 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6330419E04Rik
Genomic Map