Assay Details
| Gene Symbol: 6330436F06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018204.1 | - | 1 | 1252 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6370 | Mir6370 |
Gm27480, mmu-mir-6370 |
-1.7mb | Sense | Chr. 6 - 19555342 - 19555461 on NCBI Build GRCm38 |
| Downstream | tetraspanin 12 | Tspan12 |
9030619E17, AI426782, AI663988, AW111457, Tm4sf12 |
494.8kb | Anti-sense | Chr. 6 - 21771391 - 21853349 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6330436F06Rik
Genomic Map