Assay Details
| Gene Symbol: 6330437I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018205.1 | - | 1 | 1052 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | multiciliate differentiation and DNA synthesis associated cell cycle protein | Mcidas |
EG622408, Gm6320, Idas, Mci, Mcin |
Anti-sense | Chr. 13 - 112993472 - 113000394 on NCBI Build GRCm38 | |
| Downstream | cell division cycle 20B | Cdc20b |
EG238896, EG622422 |
33.7kb | Anti-sense | Chr. 13 - 113032954 - 113121019 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6330437I11Rik
Genomic Map